missing translation for 'onlineSavingsMsg'
Learn More

2-Pentylfuran 98.0+%, TCI America™

$117.43 - $350.44

Chemical Identifiers

CAS 3777-69-3
Molecular Formula C9H14O
Molecular Weight (g/mol) 138.21
MDL Number MFCD00036497
InChI Key YVBAUDVGOFCUSG-UHFFFAOYSA-N
Synonym furan, 2-pentyl, 2-n-pentylfuran, 2-amylfuran, furan, pentyl, pentylfuran, natural, unii-6i0qaj1jzq, 2-pentyl furan, fema no. 3317, 6i0qaj1jzq
PubChem CID 19602
IUPAC Name 2-pentylfuran
SMILES CCCCCC1=CC=CO1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
P12095ML
View Documents
TCI America
P12095ML
5 mL
Each for $117.43
Only null left
 
P120925ML
missing translation for 'viewDocuments'
TCI America
P120925ML
25 mL
Each for $350.44
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3777-69-3
138.21
YVBAUDVGOFCUSG-UHFFFAOYSA-N
19602
CCCCCC1=CC=CO1
C9H14O
MFCD00036497
furan, 2-pentyl, 2-n-pentylfuran, 2-amylfuran, furan, pentyl, pentylfuran, natural, unii-6i0qaj1jzq, 2-pentyl furan, fema no. 3317, 6i0qaj1jzq
2-pentylfuran

Specifications

3777-69-3
155°C
MFCD00036497
1993
YVBAUDVGOFCUSG-UHFFFAOYSA-N
2-pentylfuran
19602
≥98.0% (GC)
2-Pentylfuran
Yellow
C9H14O
5 mL
furan, 2-pentyl, 2-n-pentylfuran, 2-amylfuran, furan, pentyl, pentylfuran, natural, unii-6i0qaj1jzq, 2-pentyl furan, fema no. 3317, 6i0qaj1jzq
CCCCCC1=CC=CO1
138.21
138.21
Liquid
Videos
Safety and Handling

Safety and Handling

RTECSNumber : LU5187000

TSCA : Yes

SDS
Documents

Documents