missing translation for 'onlineSavingsMsg'
Learn More

2-(p-Tolyl)ethylamine, 97%

Catalog No. p-3779686
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC157880010 1 g Glass bottle
AC157880050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC157880010 Supplier Thermo Scientific Chemicals Supplier No. 157880010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3261-62-9 | C9H13N | 135.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3261-62-9
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00008195
InChI Key VKJXAQYPOTYDLO-UHFFFAOYSA-N
Synonym 4-methylphenethylamine, 2-p-tolyl ethanamine, 2-p-tolyl ethylamine, 2-p-tolyl-ethylamine, p-methylphenethylamine, benzeneethanamine, 4-methyl, 2-4-methylphenyl ethylamine, 2-4-methylphenyl ethan-1-amine, 2-4-methylphenyl ethanamine, 2-p-tolylethanamine
PubChem CID 76751
IUPAC Name 2-(4-methylphenyl)ethan-1-amine
SMILES CC1=CC=C(CCN)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9300g/mL
Boiling Point 214°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.5250 to 1.528
Linear Formula CH3C6H4CH2CH2NH2
Quantity 1 g
Beilstein 12, 1150
Specific Gravity 0.93
Formula Weight 135.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-(p-Tolyl)ethylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 221-865-2

RUO – Research Use Only