missing translation for 'onlineSavingsMsg'
Learn More

2-(p-Tolyl)benzoic Acid 98.0+%, TCI America™

Catalog No. p-7137186
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23775G 5 g
T237725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23775G Supplier TCI America Supplier No. T23775G

Chemical Identifiers

CAS 7148-03-0
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.248
MDL Number MFCD00045826
InChI Key ZSTUEICKYWFYIC-UHFFFAOYSA-N
PubChem CID 81568
IUPAC Name 2-(4-methylphenyl)benzoic acid
SMILES CC1=CC=C(C=C1)C2=CC=CC=C2C(=O)O

Specifications

Melting Point 150°C
Color White-Yellow
Quantity 5 g
Synonym 2-(4-Methylphenyl)benzoic Acid, 4′C-Methylbiphenyl-2-carboxylic Acid
Formula Weight 212.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(p-Tolyl)benzoic Acid