missing translation for 'onlineSavingsMsg'
Learn More

2-(p-Toluoyl)benzoic Acid 98.0+%, TCI America™

Catalog No. T0308500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0308500G 500 g
T030825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0308500G Supplier TCI America Supplier No. T0308500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 85-55-2
Molecular Formula C15H12O3
Molecular Weight (g/mol) 240.26
MDL Number MFCD00020287
InChI Key ICQOWIXIHDDXDI-UHFFFAOYSA-N
Synonym 2-4-methylbenzoyl benzoic acid, 2-p-toluoyl benzoic acid, benzoic acid, 2-4-methylbenzoyl, 2-p-toluyl benzoic acid, o-p-toluoyl benzoic acid, benzoic acid, o-p-toluoyl, 2-p-toluoylbenzoic acid, unii-4dn9g70pqf, 4'-methylbenzophenone-2-carboxylic acid, p-toluoyl-o-benzoic acid
PubChem CID 66563
IUPAC Name 2-(4-methylbenzoyl)benzoic acid
SMILES CC1=CC=C(C=C1)C(=O)C1=CC=CC=C1C(O)=O

Specifications

Melting Point 138°C
Color White-Yellow
Quantity 500 g
Formula Weight 240.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(p-Toluoyl)benzoic Acid
EINECSNumber (4)-0139
TSCA Yes