missing translation for 'onlineSavingsMsg'
Learn More

2-(p-Toluenesulfonyloxy)acetic Acid 97.0+%, TCI America™

Catalog No. T27271G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T27271G Supplier TCI America Supplier No. T27271G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 39794-77-9
Molecular Formula C9H10O5S
Molecular Weight (g/mol) 230.234
MDL Number MFCD00021764
InChI Key NSORSORRXHLKQV-UHFFFAOYSA-N
Synonym 2-(Tosyloxy)acetic Acid
PubChem CID 226266
IUPAC Name 2-(4-methylphenyl)sulfonyloxyacetic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCC(=O)O

Specifications

Melting Point 140°C
Color White
Quantity 1 g
Formula Weight 230.23
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(p-Toluenesulfonyloxy)acetic Acid