accessibility menu, dialog, popup

UserName

CAS RN 39794-77-9

IUPAC Name: 2-[(4-methylbenzenesulfonyl)oxy]acetic acid
Synonyms: [(4-methylbenzenesulfonyl)oxy]acetic acid
Molecular Weight (g/mol): 230.23
Molecular Formula: C9H10O5S
InChi Key: NSORSORRXHLKQV-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OCC(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results