missing translation for 'onlineSavingsMsg'
Learn More

2-(p-Toluenesulfonyl)acetic Acid 98.0+%, TCI America™

Catalog No. p-7137207
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24165G 5 g
T241625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24165G Supplier TCI America Supplier No. T24165G
Only null left

Chemical Identifiers

CAS 3937-96-0
Molecular Formula C9H10O4S
Molecular Weight (g/mol) 214.235
MDL Number MFCD00021764
InChI Key AQDHXMBUTDLAMD-UHFFFAOYSA-N
Synonym 2-tosylacetic acid, 4-toluenesulfonylacetic acid, 4-methylphenyl sulfonyl acetic acid, 2-p-toluenesulfonyl acetic acid, tosylacetic acid, unii-yid78nf77o, p-toluenesulphonylacetic acid, 4-toluenesulphonylacetic acid, p-toluenesulfonylacetic acid, yid78nf77o
PubChem CID 77538
IUPAC Name 2-(4-methylphenyl)sulfonylacetic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)CC(=O)O

Specifications

Melting Point 118°C
Quantity 5 g
Formula Weight 214.24
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(p-Toluenesulfonyl)acetic Acid