missing translation for 'onlineSavingsMsg'
Learn More

2-Oxo-2H-chromen-7-yl Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. p-7134354
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04721G 1 g
O04725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O04721G Supplier TCI America Supplier No. O04721G
Only null left

Chemical Identifiers

CAS 108530-10-5
Molecular Formula C10H5F3O5S
Molecular Weight (g/mol) 294.2
MDL Number MFCD18382236
InChI Key RHZZAJVDTLUWRD-UHFFFAOYSA-N
Synonym Trifluoromethanesulfonic Acid 2-Oxo-2H-chromen-7-yl Ester, 7-Coumaryl Trifluoromethanesulfonate, 7-Coumaryl Triflate
PubChem CID 13704479
IUPAC Name (2-oxochromen-7-yl) trifluoromethanesulfonate
SMILES C1=CC(=CC2=C1C=CC(=O)O2)OS(=O)(=O)C(F)(F)F

Specifications

Melting Point 83°C
Color White-Yellow
Quantity 1 g
Formula Weight 294.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Oxo-2H-chromen-7-yl Trifluoromethanesulfonate