accessibility menu, dialog, popup

UserName

OOOSOOFFF

CAS RN 108530-10-5

IUPAC Name: 2-oxo-2H-chromen-7-yl trifluoromethanesulfonate
Synonyms: 2-oxochromen-7-yl trifluoromethanesulfonate
Molecular Weight (g/mol): 294.2
Molecular Formula: C10H5F3O5S
InChi Key: RHZZAJVDTLUWRD-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)OC1=CC=C2C=CC(=O)OC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results