missing translation for 'onlineSavingsMsg'
Learn More

2-Oxazolidone 98.0+%, TCI America™

Catalog No. p-7134232
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O018825G 25 g
O0188100G 100 g
O0188500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O018825G Supplier TCI America Supplier No. O018825G
Only null left

Chemical Identifiers

CAS 497-25-6
Molecular Formula C3H5NO2
Molecular Weight (g/mol) 87.08
MDL Number MFCD00005268
InChI Key IZXIZTKNFFYFOF-UHFFFAOYSA-N
Synonym 2-oxazolidone, oxazolidin-2-one, 2-oxazolidinone, oxazolidinone, oxazolidone, oxazolidinones, unii-z4d49w92pp, carbamic acid, 2-hydroxyethyl-, gamma-lactone, wln: t5mvotj, carbamic acid, .gamma.-lactone
PubChem CID 73949
ChEBI CHEBI:1237
IUPAC Name 1,3-oxazolidin-2-one
SMILES O=C1NCCO1

Specifications

Melting Point 89°C
Color White-Yellow
Boiling Point 200°C
Quantity 25 g
Formula Weight 87.08
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Oxazolidone
RTECSNumber RQ2450000
TSCA Yes