accessibility menu, dialog, popup

UserName

ONHO

CAS RN 497-25-6

IUPAC Name: 1,3-oxazolidin-2-one
Synonyms: oxazolidinone
Molecular Weight (g/mol): 87.08
Molecular Formula: C3H5NO2
InChi Key: IZXIZTKNFFYFOF-UHFFFAOYSA-N
SMILES: O=C1NCCO1
Boiling Point 
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (3)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)