missing translation for 'onlineSavingsMsg'
Learn More

2-Oxazolidinone, 99%

CAS: 497-25-6 | C3H5NO2 | 87.08 g/mol

Supplier:  Thermo Scientific Chemicals A1454322

 View more versions of this product

Catalog No. AAA1454322

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
2-Oxazolidinone is used as antimicrobials, in organisms it act as a protein synthesis inhibitors, targeting an early step involving the binding of N-formylmethionyl-tRNA to the ribosome. They are used for chiral synthesis. It has been used as an ethanolamine equivalent, also behaves as an aziridine equivalent as in the conversion of thiophenols or amines to their aminoethyl derivatives.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

2-Oxazolidinone
84°C to 89°C
C3H5NO2
100 g
2-oxazolidone, oxazolidin-2-one, 2-oxazolidinone, oxazolidinone, oxazolidone, oxazolidinones, unii-z4d49w92pp, carbamic acid, 2-hydroxyethyl-, gamma-lactone, wln: t5mvotj, carbamic acid, .gamma.-lactone
IZXIZTKNFFYFOF-UHFFFAOYSA-N
1,3-oxazolidin-2-one
73949
87.08
497-25-6
220°C (48 mmHg)
MFCD00005268
106251
Soluble in water.
O=C1NCCO1
87.08
CHEBI:1237
99%

Chemical Identifiers

497-25-6
87.08
IZXIZTKNFFYFOF-UHFFFAOYSA-N
73949
1,3-oxazolidin-2-one
C3H5NO2
MFCD00005268
2-oxazolidone, oxazolidin-2-one, 2-oxazolidinone, oxazolidinone, oxazolidone, oxazolidinones, unii-z4d49w92pp, carbamic acid, 2-hydroxyethyl-, gamma-lactone, wln: t5mvotj, carbamic acid, .gamma.-lactone
CHEBI:1237
O=C1NCCO1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 207-840-9

RTECSNumber : RQ2450000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only