missing translation for 'onlineSavingsMsg'
Learn More

2-Oxazolidinone, 99%

Catalog No. AAA1454336
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1454336 500 g
AAA1454314 25 g
AAA1454322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1454336 Supplier Thermo Scientific Chemicals Supplier No. A1454336
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 497-25-6 | C3H5NO2 | 87.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
2-Oxazolidinone is used as antimicrobials, in organisms it act as a protein synthesis inhibitors, targeting an early step involving the binding of N-formylmethionyl-tRNA to the ribosome. They are used for chiral synthesis. It has been used as an ethanolamine equivalent, also behaves as an aziridine equivalent as in the conversion of thiophenols or amines to their aminoethyl derivatives.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 497-25-6
Molecular Formula C3H5NO2
Molecular Weight (g/mol) 87.08
MDL Number MFCD00005268
InChI Key IZXIZTKNFFYFOF-UHFFFAOYSA-N
Synonym 2-oxazolidone, oxazolidin-2-one, 2-oxazolidinone, oxazolidinone, oxazolidone, oxazolidinones, unii-z4d49w92pp, carbamic acid, 2-hydroxyethyl-, gamma-lactone, wln: t5mvotj, carbamic acid, .gamma.-lactone
PubChem CID 73949
ChEBI CHEBI:1237
IUPAC Name 1,3-oxazolidin-2-one
SMILES O=C1NCCO1

Specifications

Melting Point 84°C to 89°C
Boiling Point 220°C (48 mmHg)
Quantity 500 g
Beilstein 106251
Solubility Information Soluble in water.
Formula Weight 87.08
Percent Purity 99%
Chemical Name or Material 2-Oxazolidinone
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 207-840-9
RTECSNumber RQ2450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only