missing translation for 'onlineSavingsMsg'
Learn More

2-Oxa-6-azaspiro[3.3]heptane 98.0+%, TCI America™

Catalog No. O04661G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O04661G Supplier TCI America Supplier No. O04661G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 174-78-7
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.133
MDL Number MFCD12407093
InChI Key HPJALMWOZYIZGE-UHFFFAOYSA-N
PubChem CID 23111887
IUPAC Name 2-oxa-6-azaspiro[3.3]heptane
SMILES C1C2(CN1)COC2

Specifications

Color Yellow
Quantity 1 g
UN Number 1993
Formula Weight 99.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Oxa-6-azaspiro[3.3]heptane
TSCA No
Recommended Storage Freezer