accessibility menu, dialog, popup

UserName

HNO

CAS RN 174-78-7

IUPAC Name: 2-oxa-6-azaspiro[3.3]heptane
Synonyms: 2-oxa-6-azaspiro[3.3]heptane
Molecular Weight (g/mol): 99.13
Molecular Formula: C5H9NO
InChi Key: HPJALMWOZYIZGE-UHFFFAOYSA-N
SMILES: C1NCC11COC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results