missing translation for 'onlineSavingsMsg'
Learn More

2-Octyne, 97%, stabilized, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 455570250

 View more versions of this product

Catalog No. AC455570250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

2-Octyne
0.7590g/mL
18°C
96% min. (GC)
MFCD00027302
0.759
QCQALVMFTWRCFI-UHFFFAOYSA-N
oct-2-yne
110.20
110.2
2809-67-8
137.0°C
Authentic
C8H14
25g
2-octyne, n-c5h11c.$.cch3, amylmethylacetylene, methylpentylacetylene, acmc-209h0u, n-c5h11c.equiv.cch3
CCCCCC#CC
Approx. 200ppm
17769
97%

Chemical Identifiers

2809-67-8
110.20
QCQALVMFTWRCFI-UHFFFAOYSA-N
17769
CCCCCC#CC
C8H14
MFCD00027302
2-octyne, n-c5h11c.$.cch3, amylmethylacetylene, methylpentylacetylene, acmc-209h0u, n-c5h11c.equiv.cch3
oct-2-yne
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea

GHS Signal Word: Danger

EINECSNumber : 220-553-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only