missing translation for 'onlineSavingsMsg'
Learn More

2-Octyne, 97%, stabilized, Thermo Scientific™

Catalog No. p-7077564
Change view
Click to view available options
Quantity:
5g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455570050 5g
AC455570250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC455570050 Supplier Thermo Scientific Chemicals Supplier No. 455570050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2809-67-8
Molecular Formula C8H14
Molecular Weight (g/mol) 110.20
MDL Number MFCD00027302
InChI Key QCQALVMFTWRCFI-UHFFFAOYSA-N
Synonym 2-octyne, n-c5h11c.$.cch3, amylmethylacetylene, methylpentylacetylene, acmc-209h0u, n-c5h11c.equiv.cch3
PubChem CID 17769
IUPAC Name oct-2-yne
SMILES CCCCCC#CC

Specifications

Density 0.7590g/mL
Boiling Point 137.0°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5g
Specific Gravity 0.759
Stabilizer Approx. 200ppm
Formula Weight 110.2
Percent Purity 97%
Chemical Name or Material 2-Octyne
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea
EINECSNumber 220-553-3

RUO – Research Use Only