missing translation for 'onlineSavingsMsg'
Learn More

2-Octenylsuccinic Anhydride (cis- and trans- mixture) 95.0+%, TCI America™

$70.84 - $493.97

Chemical Identifiers

CAS 42482-06-4
Molecular Formula C12H18O3
Molecular Weight (g/mol) 210.273
MDL Number MFCD00047133
InChI Key WSGFXVFLWVXTCJ-VOTSOKGWSA-N
PubChem CID 5362689
IUPAC Name 3-[(E)-oct-2-enyl]oxolane-2,5-dione
SMILES CCCCCC=CCC1CC(=O)OC1=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
O004025G
View Documents
TCI America
O004025G
25 g N/A
Only null left
 
O0040500G
View Documents
TCI America
O0040500G
500 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

42482-06-4
210.273
WSGFXVFLWVXTCJ-VOTSOKGWSA-N
3-[(E)-oct-2-enyl]oxolane-2,5-dione
C12H18O3
MFCD00047133
5362689
CCCCCC=CCC1CC(=O)OC1=O

Specifications

42482-06-4
25 g
C12H18O3
WSGFXVFLWVXTCJ-VOTSOKGWSA-N
3-[(E)-oct-2-enyl]oxolane-2,5-dione
5362689
≥95.0% (GC,T)
Yellow
Clear Liquid at 20°C
MFCD00047133
CCCCCC=CCC1CC(=O)OC1=O
210.273
210.27
2-Octenylsuccinic Anhydride (cis- and trans- mixture)
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0852

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'