missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Octenylsuccinic Anhydride (cis- and trans- mixture) 95.0+%, TCI America™
Supplier: TCI America O0040500G
Specifications
| 2-Octenylsuccinic Anhydride (cis- and trans- mixture) | |
| Yellow | |
| Clear Liquid at 20°C | |
| MFCD00047133 | |
| CCCCCC=CCC1CC(=O)OC1=O | |
| 210.273 | |
| 210.27 |
| 42482-06-4 | |
| 500 g | |
| C12H18O3 | |
| WSGFXVFLWVXTCJ-VOTSOKGWSA-N | |
| 3-[(E)-oct-2-enyl]oxolane-2,5-dione | |
| 5362689 | |
| ≥95.0% (GC,T) |
Chemical Identifiers
| 42482-06-4 | |
| 210.273 | |
| WSGFXVFLWVXTCJ-VOTSOKGWSA-N | |
| 3-[(E)-oct-2-enyl]oxolane-2,5-dione |
| C12H18O3 | |
| MFCD00047133 | |
| 5362689 | |
| CCCCCC=CCC1CC(=O)OC1=O |
Safety and Handling
EINECSNumber : (2)-0852
TSCA : Yes