missing translation for 'onlineSavingsMsg'
Learn More

2-Octanone 98.0+%, TCI America™

Catalog No. O003825ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O003825ML 25 mL
O0038500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O003825ML Supplier TCI America Supplier No. O003825ML

Chemical Identifiers

CAS 111-13-7
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00009540
InChI Key ZPVFWPFBNIEHGJ-UHFFFAOYSA-N
Synonym 2-octanone, n-hexyl methyl ketone, hexyl methyl ketone, methyl hexyl ketone, methyl n-hexyl ketone, octanone, 2-oxooctane, 2-octanone natural, fema number 2802, unii-j2g84h29af
PubChem CID 8093
ChEBI CHEBI:87434
IUPAC Name octan-2-one
SMILES CCCCCCC(=O)C

Specifications

Color Yellow
Boiling Point 173°C
Quantity 25 mL
UN Number 1224
Formula Weight 128.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Octanone
EINECSNumber (2)-0542
RTECSNumber RH1484000
TSCA Yes