missing translation for 'onlineSavingsMsg'
Learn More

2-Octanone, 98%

Catalog No. p-7081607
Change view
Click to view available options
Quantity:
250 mL
5000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA11643AK 250 mL
AAA116430J 5000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA11643AK Supplier Thermo Scientific Chemicals Supplier No. A11643AK

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 111-13-7 | C8H16O | 128.215 g/mol

2-Octanone is used in perfumes, high-boiling solvent, especially for epoxy resin coatings and in leather finishes. It is used in flavoring, odorant. It can act as antiblushing agent for nitrocellulose lacquers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Octanone is used in perfumes, high-boiling solvent, especially for epoxy resin coatings and in leather finishes. It is used in flavoring, odorant. It can act as antiblushing agent for nitrocellulose lacquers.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.

Chemical Identifiers

CAS 111-13-7
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00009540
InChI Key ZPVFWPFBNIEHGJ-UHFFFAOYSA-N
Synonym 2-octanone, n-hexyl methyl ketone, hexyl methyl ketone, methyl hexyl ketone, methyl n-hexyl ketone, octanone, 2-oxooctane, 2-octanone natural, fema number 2802, unii-j2g84h29af
PubChem CID 8093
ChEBI CHEBI:87434
IUPAC Name octan-2-one
SMILES CCCCCCC(=O)C

Specifications

Melting Point -16°C
Density 0.819
Boiling Point 172°C to 173°C
Flash Point 54°C (129°F)
Refractive Index 1.415
Quantity 250 mL
Beilstein 635843
Merck Index 14,4711
Solubility Information Slightly soluble in water.
Formula Weight 128.22
Percent Purity 98%
Chemical Name or Material 2-Octanone
Show More Show Less
Hazard Category H226-H312
Hazard Statement GHS H Statement
H311-H227-H315-H319-H335
Toxic in contact with skin.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P312-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 203-837-1
RTECSNumber RH1484000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only