missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-O-Methyluridine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 161°C |
| Color | White |
| Quantity | 1 g |
| Formula Weight | 258.23 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2′-O-Methyluridine |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |