missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-O-Methylguanosine Hydrate 98.0+%, TCI America™
$149.13 - $469.75
Chemical Identifiers
| CAS | 2140-71-8 |
|---|---|
| Molecular Formula | C11H15N5O5 |
| Molecular Weight (g/mol) | 297.27 |
| MDL Number | MFCD00057053 |
| InChI Key | OVYNGSFVYRPRCG-YPLCUDRINA-N |
| Synonym | 2'-o-methylguanosine, 2'-o-methyl guanosine, 2'-o-methyl-guanosine, guanosine, 2'-o-methyl, unii-w722h4pa1s, 2-amino-9-2-o-methyl-beta-d-ribofuranosyl-1,9-dihydro-6h-purin-6-one, 2-amino-9-2r,3r,4r,5r-4-hydroxy-5-hydroxymethyl-3-methoxyoxolan-2-yl-3h-purin-6-one, 2-amino-9-2r,3r,4r,5r-4-hydroxy-5-hydroxymethyl-3-methoxytetrahydrofuran-2-yl-1,9-dihydro-6h-purin-6-one, 2-o-methylguanosine |
| PubChem CID | 188959 |
| ChEBI | CHEBI:19229 |
| IUPAC Name | 2-amino-9-[(2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxyoxolan-2-yl]-6,9-dihydro-1H-purin-6-one |
| SMILES | CO[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C1N=C(N)NC2=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M2318200MG
|
TCI America
M2318200MG |
200 mg |
Each for $149.13
|
|
|||||
|
M23181G
|
TCI America
M23181G |
1 g |
Each for $469.75
|
|
|||||
Chemical Identifiers
| 2140-71-8 | |
| 297.27 | |
| OVYNGSFVYRPRCG-YPLCUDRINA-N | |
| 188959 | |
| 2-amino-9-[(2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxyoxolan-2-yl]-6,9-dihydro-1H-purin-6-one |
| C11H15N5O5 | |
| MFCD00057053 | |
| 2'-o-methylguanosine, 2'-o-methyl guanosine, 2'-o-methyl-guanosine, guanosine, 2'-o-methyl, unii-w722h4pa1s, 2-amino-9-2-o-methyl-beta-d-ribofuranosyl-1,9-dihydro-6h-purin-6-one, 2-amino-9-2r,3r,4r,5r-4-hydroxy-5-hydroxymethyl-3-methoxyoxolan-2-yl-3h-purin-6-one, 2-amino-9-2r,3r,4r,5r-4-hydroxy-5-hydroxymethyl-3-methoxytetrahydrofuran-2-yl-1,9-dihydro-6h-purin-6-one, 2-o-methylguanosine | |
| CHEBI:19229 | |
| CO[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C1N=C(N)NC2=O |
Specifications
| 2140-71-8 | |
| White | |
| MFCD00057053 | |
| 2'-o-methylguanosine, 2'-o-methyl guanosine, 2'-o-methyl-guanosine, guanosine, 2'-o-methyl, unii-w722h4pa1s, 2-amino-9-2-o-methyl-beta-d-ribofuranosyl-1,9-dihydro-6h-purin-6-one, 2-amino-9-2r,3r,4r,5r-4-hydroxy-5-hydroxymethyl-3-methoxyoxolan-2-yl-3h-purin-6-one, 2-amino-9-2r,3r,4r,5r-4-hydroxy-5-hydroxymethyl-3-methoxytetrahydrofuran-2-yl-1,9-dihydro-6h-purin-6-one, 2-o-methylguanosine | |
| CO[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C1N=C(N)NC2=O | |
| 297.27 | |
| CHEBI:19229 | |
| ≥98.0% (HPLC,T) | |
| 2′-O-Methylguanosine Hydrate |
| 236°C | |
| C11H15N5O5 | |
| 200 mg | |
| OVYNGSFVYRPRCG-YPLCUDRINA-N | |
| 2-amino-9-[(2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxyoxolan-2-yl]-6,9-dihydro-1H-purin-6-one | |
| 188959 | |
| 297.27 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator