accessibility menu, dialog, popup

UserName

CAS RN 2140-71-8

IUPAC Name: 2-amino-9-[(2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxyoxolan-2-yl]-6,9-dihydro-1H-purin-6-one
Synonyms: 2'-O-methylguanosine
Molecular Weight (g/mol): 297.27
Molecular Formula: C11H15N5O5
InChi Key: OVYNGSFVYRPRCG-YPLCUDRINA-N
SMILES: CO[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C1N=C(N)NC2=O
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)