missing translation for 'onlineSavingsMsg'
Learn More

2'-O-Methylguanosine, 99%, Thermo Scientific Chemicals

$147.77 - $147.77

Chemical Identifiers

CAS 2140-71-8
Molecular Formula C11H15N5O5
Molecular Weight (g/mol) 297.27
MDL Number MFCD00057053
InChI Key OVYNGSFVYRPRCG-YPLCUDRINA-N
Synonym 2'-o-methylguanosine, 2'-o-methyl guanosine, 2'-o-methyl-guanosine, guanosine, 2'-o-methyl, unii-w722h4pa1s, 2-amino-9-2-o-methyl-beta-d-ribofuranosyl-1,9-dihydro-6h-purin-6-one, 2-amino-9-2r,3r,4r,5r-4-hydroxy-5-hydroxymethyl-3-methoxyoxolan-2-yl-3h-purin-6-one, 2-amino-9-2r,3r,4r,5r-4-hydroxy-5-hydroxymethyl-3-methoxytetrahydrofuran-2-yl-1,9-dihydro-6h-purin-6-one, 2-o-methylguanosine
PubChem CID 188959
ChEBI CHEBI:19229
SMILES CO[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C1N=C(N)NC2=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6577003
View Documents
Thermo Scientific Chemicals
J6577003
1 g
Each for $147.77
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2140-71-8
297.27
OVYNGSFVYRPRCG-YPLCUDRINA-N
188959
CO[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C1N=C(N)NC2=O
C11H15N5O5
MFCD00057053
2'-o-methylguanosine, 2'-o-methyl guanosine, 2'-o-methyl-guanosine, guanosine, 2'-o-methyl, unii-w722h4pa1s, 2-amino-9-2-o-methyl-beta-d-ribofuranosyl-1,9-dihydro-6h-purin-6-one, 2-amino-9-2r,3r,4r,5r-4-hydroxy-5-hydroxymethyl-3-methoxyoxolan-2-yl-3h-purin-6-one, 2-amino-9-2r,3r,4r,5r-4-hydroxy-5-hydroxymethyl-3-methoxytetrahydrofuran-2-yl-1,9-dihydro-6h-purin-6-one, 2-o-methylguanosine
CHEBI:19229

Specifications

2140-71-8
C11H15N5O5
1 g
OVYNGSFVYRPRCG-YPLCUDRINA-N
2-amino-9-[(2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxyoxolan-2-yl]-3H-purin-6-one
188959
297.27
Powder
White
MFCD00057053
2'-o-methylguanosine, 2'-o-methyl guanosine, 2'-o-methyl-guanosine, guanosine, 2'-o-methyl, unii-w722h4pa1s, 2-amino-9-2-o-methyl-beta-d-ribofuranosyl-1,9-dihydro-6h-purin-6-one, 2-amino-9-2r,3r,4r,5r-4-hydroxy-5-hydroxymethyl-3-methoxyoxolan-2-yl-3h-purin-6-one, 2-amino-9-2r,3r,4r,5r-4-hydroxy-5-hydroxymethyl-3-methoxytetrahydrofuran-2-yl-1,9-dihydro-6h-purin-6-one, 2-o-methylguanosine
CO[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C1N=C(N)NC2=O
297.27
CHEBI:19229
99%
2'-O-Methylguanosine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only