missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-O-alpha-D-Glucopyranosyl-L-ascorbic Acid 98.0+%, TCI America™
$49.36 - $146.20
Chemical Identifiers
| CAS | 129499-78-1 |
|---|---|
| Molecular Formula | C12H18O11 |
| Molecular Weight (g/mol) | 338.265 |
| MDL Number | MFCD00925623 |
| InChI Key | MLSJBGYKDYSOAE-DCWMUDTNSA-N |
| Synonym | l-ascorbic acid 2-glucoside, aa-2g, unii-2v52r0nhxw, 2-o-alpha-d-glucopyranosyl-l-ascorbic acid, 2v52r0nhxw, l-ascorbic acid-2-glucoside, ascorbic acid 2-o-glucoside, ascorbyl glucoside, l-ascorbic acid 2-o-alpha-glucoside, l-ascorbicacid2-glucoside |
| PubChem CID | 54693473 |
| ChEBI | CHEBI:81685 |
| IUPAC Name | (2R)-2-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-4-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2H-furan-5-one |
| SMILES | C(C1C(C(C(C(O1)OC2=C(C(OC2=O)C(CO)O)O)O)O)O)O |
Chemical Identifiers
| 129499-78-1 | |
| 338.265 | |
| MLSJBGYKDYSOAE-DCWMUDTNSA-N | |
| 54693473 | |
| (2R)-2-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-4-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2H-furan-5-one |
| C12H18O11 | |
| MFCD00925623 | |
| l-ascorbic acid 2-glucoside, aa-2g, unii-2v52r0nhxw, 2-o-alpha-d-glucopyranosyl-l-ascorbic acid, 2v52r0nhxw, l-ascorbic acid-2-glucoside, ascorbic acid 2-o-glucoside, ascorbyl glucoside, l-ascorbic acid 2-o-alpha-glucoside, l-ascorbicacid2-glucoside | |
| CHEBI:81685 | |
| C(C1C(C(C(C(O1)OC2=C(C(OC2=O)C(CO)O)O)O)O)O)O |
Specifications
| 129499-78-1 | |
| White | |
| MFCD00925623 | |
| l-ascorbic acid 2-glucoside, aa-2g, unii-2v52r0nhxw, 2-o-alpha-d-glucopyranosyl-l-ascorbic acid, 2v52r0nhxw, l-ascorbic acid-2-glucoside, ascorbic acid 2-o-glucoside, ascorbyl glucoside, l-ascorbic acid 2-o-alpha-glucoside, l-ascorbicacid2-glucoside | |
| C(C1C(C(C(C(O1)OC2=C(C(OC2=O)C(CO)O)O)O)O)O)O | |
| 338.265 | |
| CHEBI:81685 | |
| ≥98.0% (HPLC,T) | |
| 2-O-alpha-D-Glucopyranosyl-L-ascorbic Acid |
| 160°C | |
| C12H18O11 | |
| 1 g | |
| MLSJBGYKDYSOAE-DCWMUDTNSA-N | |
| (2R)-2-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-4-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2H-furan-5-one | |
| 54693473 | |
| 338.27 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-6596
TSCA : No