missing translation for 'onlineSavingsMsg'
Learn More

2-O-alpha-D-Glucopyranosyl-L-ascorbic Acid 98.0+%, TCI America™

Catalog No. G03941G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G03941G 1 g
G03945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G03941G Supplier TCI America Supplier No. G03941G

Chemical Identifiers

CAS 129499-78-1
Molecular Formula C12H18O11
Molecular Weight (g/mol) 338.265
MDL Number MFCD00925623
InChI Key MLSJBGYKDYSOAE-DCWMUDTNSA-N
Synonym l-ascorbic acid 2-glucoside, aa-2g, unii-2v52r0nhxw, 2-o-alpha-d-glucopyranosyl-l-ascorbic acid, 2v52r0nhxw, l-ascorbic acid-2-glucoside, ascorbic acid 2-o-glucoside, ascorbyl glucoside, l-ascorbic acid 2-o-alpha-glucoside, l-ascorbicacid2-glucoside
PubChem CID 54693473
ChEBI CHEBI:81685
IUPAC Name (2R)-2-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-4-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2H-furan-5-one
SMILES C(C1C(C(C(C(O1)OC2=C(C(OC2=O)C(CO)O)O)O)O)O)O

Specifications

Melting Point 160°C
Color White
Quantity 1 g
Formula Weight 338.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-O-alpha-D-Glucopyranosyl-L-ascorbic Acid
EINECSNumber (5)-6596
TSCA No