missing translation for 'onlineSavingsMsg'
Learn More

2-O-alpha-D-Glucopyranosyl-L-ascorbic acid, 98+%

Catalog No. AAJ6660103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6660103 1 g
AAJ6660106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6660103 Supplier Thermo Scientific Chemicals Supplier No. J6660103

CAS: 129499-78-1 | C12H18O11 | 338.265 g/mol

2-O-alpha-D-Glucopyranosyl-L-ascorbic acid reduce the free radicals that result from UV irradiation of the skin, and significantly reduce cell damage and photo-aging. It also promotes collagen synthesis. It can function as an antioxidant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-O-alpha-D-Glucopyranosyl-L-ascorbic acid reduce the free radicals that result from UV irradiation of the skin, and significantly reduce cell damage and photo-aging. It also promotes collagen synthesis. It can function as an antioxidant.

Solubility
Soluble in water. (879 g/L) at 25°C.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.

Chemical Identifiers

CAS 129499-78-1
Molecular Formula C12H18O11
Molecular Weight (g/mol) 338.265
MDL Number MFCD23701380
InChI Key MLSJBGYKDYSOAE-DCWMUDTNSA-N
Synonym l-ascorbic acid 2-glucoside, aa-2g, unii-2v52r0nhxw, 2-o-alpha-d-glucopyranosyl-l-ascorbic acid, 2v52r0nhxw, l-ascorbic acid-2-glucoside, ascorbic acid 2-o-glucoside, ascorbyl glucoside, l-ascorbic acid 2-o-alpha-glucoside, l-ascorbicacid2-glucoside
PubChem CID 54693473
ChEBI CHEBI:81685
IUPAC Name (2R)-2-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-4-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2H-furan-5-one
SMILES C(C1C(C(C(C(O1)OC2=C(C(OC2=O)C(CO)O)O)O)O)O)O

Specifications

Melting Point 158°C to 163°C
Color White
Quantity 1 g
Solubility Information Soluble in water. (879g/L) at 25°C.
Formula Weight 338.26
Percent Purity 99.5%
Physical Form Powder
Chemical Name or Material 2-O-alpha-D-Glucopyranosyl-L-ascorbic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only