missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Nonylbenzimidazole 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 127°C |
| Color | White |
| Boiling Point | 280°C |
| Quantity | 25 g |
| Formula Weight | 244.38 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Nonylbenzimidazole |
Safety and Handling
| TSCA | No |