accessibility menu, dialog, popup

UserName

CAS RN 5851-50-3

H3CNHN

CAS RN 5851-50-3

IUPAC Name: 2-nonyl-1H-1,3-benzodiazole
Synonyms: 2-nonyl-1H-1,3-benzodiazole
Molecular Weight (g/mol): 244.38
Molecular Formula: C16H24N2
InChi Key: YECSLYXTXWSKBO-UHFFFAOYSA-N
SMILES: CCCCCCCCCC1=NC2=CC=CC=C2N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results