missing translation for 'onlineSavingsMsg'
Learn More

2-Nonadecanone, tech. 80%

Catalog No. AAB2117806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2117806 5 g
AAB2117814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2117806 Supplier Thermo Scientific Chemicals Supplier No. B2117806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 629-66-3 | C19H38O | 282.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 629-66-3
Molecular Formula C19H38O
Molecular Weight (g/mol) 282.51
MDL Number MFCD00026671
InChI Key IEDKVDCIEARIIU-UHFFFAOYSA-N
Synonym 2-nonadecanone, nonadeca-2-one, methyl heptadecyl ketone, acmc-20anop, 2-oxononadecan-3-yl, n-heptadecyl methyl ketone
PubChem CID 69423
IUPAC Name nonadecan-2-one
SMILES CCCCCCCCCCCCCCCCCC(C)=O

Specifications

Melting Point 54°C to 56°C
Flash Point >100°C (212°F)
Linear Formula CH3(CH2)16COCH3
Quantity 5 g
Beilstein 1708703
Formula Weight 282.51
Percent Purity 80%
Grade Technical
Chemical Name or Material 2-Nonadecanone
EINECSNumber 211-102-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only