accessibility menu, dialog, popup

UserName

CAS RN 629-66-3

IUPAC Name: nonadecan-2-one
Synonyms: 2-nonadecanone
Molecular Weight (g/mol): 282.51
Molecular Formula: C19H38O
InChi Key: IEDKVDCIEARIIU-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC(C)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)