missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrothiophene, tech. 85%

Catalog No. p-7023666
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1746406 5 g
AAA1746414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1746406 Supplier Thermo Scientific Chemicals Supplier No. A1746406
Only null left

CAS: 609-40-5 | C4H3NO2S | 129.133 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 609-40-5
Molecular Formula C4H3NO2S
Molecular Weight (g/mol) 129.133
MDL Number MFCD00005425
InChI Key JIZRGGUCOQKGQD-UHFFFAOYSA-N
Synonym thiophene, 2-nitro, 2-nitro-thiophene, 2-nitro thiophene, 2-nitrothiophene, tech, nitrothiophene, 2-nitrothiofene, pubchem5570, 2-nitrothiophene, tech., acmc-1aw2t, wln: t5sj bnw
PubChem CID 11866
IUPAC Name 2-nitrothiophene
SMILES C1=CSC(=C1)[N+](=O)[O-]

Specifications

Melting Point 43°C to 45°C
Density 1.3644
Boiling Point 224°C to 225°C
Flash Point 94°C (201°F)
Quantity 5 g
Sensitivity Light sensitive
Formula Weight 129.14
Percent Purity 85%
Grade Technical
Chemical Name or Material 2-Nitrothiophene
Hazard Category H302+H312+H332-H341
Hazard Statement GHS H Statement
H341
Suspected of causing genetic defects.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
EINECSNumber 210-190-9
RTECSNumber XN0030000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only