accessibility menu, dialog, popup

UserName

ONOS

CAS RN 609-40-5

IUPAC Name: 2-nitrothiophene
Synonyms: 2-nitrothiophene
Molecular Weight (g/mol): 129.13
Molecular Formula: C4H3NO2S
InChi Key: JIZRGGUCOQKGQD-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CS1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)