missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroterephthalic acid 4-methyl ester, 97%

Catalog No. AAH2777914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2777914 25 g
AAH2777906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2777914 Supplier Thermo Scientific Chemicals Supplier No. H2777914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 55737-66-1 | C9H7NO6 | 225.156 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 55737-66-1
Molecular Formula C9H7NO6
Molecular Weight (g/mol) 225.156
MDL Number MFCD06203344
InChI Key VULISSQANNKDCH-UHFFFAOYSA-N
Synonym 4-methoxycarbonyl-2-nitrobenzoic acid, 2-nitro-4-methoxycarbonyl benzoic acid, 2-nitroterephthalic acid 4-methyl ester, acmc-1awlg, methyl 4-carboxy-3-nitrobenzoate, 4-carbomethoxy-2-nitrobenzoic acid, 4-methoxycarbonyl-2-nitrobenzoicacid, 2-nitro-terephthalic acid 4-methyl ester
PubChem CID 21906474
SMILES COC(=O)C1=CC(=C(C=C1)C(=O)O)[N+](=O)[O-]

Specifications

Melting Point 132°C to 136°C
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 225.17
Percent Purity 97%
Chemical Name or Material 2-Nitroterephthalic acid 4-methyl ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No

RUO – Research Use Only