missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroresorcinol, 98%

Catalog No. AC416160100
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC416160100 10 g Glass bottle
AC416160250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC416160100 Supplier Thermo Scientific Chemicals Supplier No. 416160100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 601-89-8 | C6H5NO4 | 155.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 601-89-8
Molecular Formula C6H5NO4
Molecular Weight (g/mol) 155.11
MDL Number MFCD00007124
InChI Key ZLCPKMIJYMHZMJ-UHFFFAOYSA-N
Synonym 2-nitroresorcinol, 2-nitrobenzene-1,3-diol, 1,3-benzenediol, 2-nitro, resorcinol, 2-nitro, 2-nitro-1,3-benzenediol, 1,3-dihydroxy-2-nitrobenzene, 1,3-benzenediol, nitro, 2,6-dihydroxynitrobenzene, nitroresorcinol, acmc-209mid
PubChem CID 11760
IUPAC Name 2-nitrobenzene-1,3-diol
SMILES OC1=CC=CC(O)=C1[N+]([O-])=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 80°C to 84°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 10 g
Formula Weight 155.11
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitroresorcinol, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent
EINECSNumber 210-010-9
TSCA TSCA

RUO – Research Use Only