missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroresorcinol, 98%

Catalog No. AAA1297022
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1297022 100 g
AAA1297006 5 g
AAA1297014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1297022 Supplier Thermo Scientific Chemicals Supplier No. A1297022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 601-89-8 | C6H5NO4 | 155.11 g/mol

It is used in the synthesis of porous materials by 2-nitroresorcinol/cyanuric chloride thermal polycondensation in emulsions. It is used as an active pharmaceutical intermediate and an intermediate for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of porous materials by 2-nitroresorcinol/cyanuric chloride thermal polycondensation in emulsions. It is used as an active pharmaceutical intermediate and an intermediate for organic synthesis.

Solubility
Slightly soluble in water. Solubility in methanol is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 601-89-8
Molecular Formula C6H5NO4
Molecular Weight (g/mol) 155.11
MDL Number MFCD00007124
InChI Key ZLCPKMIJYMHZMJ-UHFFFAOYSA-N
Synonym 2-nitroresorcinol, 2-nitrobenzene-1,3-diol, 1,3-benzenediol, 2-nitro, resorcinol, 2-nitro, 2-nitro-1,3-benzenediol, 1,3-dihydroxy-2-nitrobenzene, 1,3-benzenediol, nitro, 2,6-dihydroxynitrobenzene, nitroresorcinol, acmc-209mid
PubChem CID 11760
IUPAC Name 2-nitrobenzene-1,3-diol
SMILES OC1=CC=CC(O)=C1[N+]([O-])=O

Specifications

Melting Point 81°C to 85°C
Quantity 100 g
UN Number UN2811
Beilstein 2048819
Solubility Information Slightly soluble in water. Solubility in methanol is almost transparent.
Formula Weight 155.11
Percent Purity 98%
Chemical Name or Material 2-Nitroresorcinol
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 210-010-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only