missing translation for 'onlineSavingsMsg'
Learn More

2-Nitropropane 95.0+%, TCI America™

$208.78 - $285.21

Chemical Identifiers

CAS 79-46-9
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.094
MDL Number MFCD00007397
InChI Key FGLBSLMDCBOPQK-UHFFFAOYSA-N
Synonym propane, 2-nitro, isonitropropane, dimethylnitromethane, nitroisopropane, sec-nitropropane, nipar s-20, beta-nitropropane, 2-np, nipar s-20 solvent, nipar s-30 solvent
PubChem CID 398
ChEBI CHEBI:16037
IUPAC Name 2-nitropropane
SMILES CC(C)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N024925G
View Documents
TCI America
N024925G
25 g
Each for $208.78
Only null left
 
N0249100G
View Documents
TCI America
N0249100G
100 g
Each for $285.21
Only null left
 
Description

Description

Specifications

Chemical Identifiers

79-46-9
89.094
FGLBSLMDCBOPQK-UHFFFAOYSA-N
398
2-nitropropane
C3H7NO2
MFCD00007397
propane, 2-nitro, isonitropropane, dimethylnitromethane, nitroisopropane, sec-nitropropane, nipar s-20, beta-nitropropane, 2-np, nipar s-20 solvent, nipar s-30 solvent
CHEBI:16037
CC(C)[N+](=O)[O-]

Specifications

79-46-9
Yellow
C3H7NO2
25 g
propane, 2-nitro, isonitropropane, dimethylnitromethane, nitroisopropane, sec-nitropropane, nipar s-20, beta-nitropropane, 2-np, nipar s-20 solvent, nipar s-30 solvent
CC(C)[N+](=O)[O-]
89.094
CHEBI:16037
≥95.0% (GC)
2-Nitropropane
-91°C
120°C
MFCD00007397
2608
FGLBSLMDCBOPQK-UHFFFAOYSA-N
2-nitropropane
398
89.09
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0194

RTECSNumber : TZ5250000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'