missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Nitrophenylsulfenyl Chloride 95.0+%, TCI America™
Supplier: TCI America N036325G
Specifications
| 2-Nitrophenylsulfenyl Chloride [N-Protecting Agent for Peptides Research] | |
| 75°C | |
| C6H4ClNO2S | |
| 25 g | |
| 2-nitrobenzenesulfenyl chloride, 2-nitrophenylsulfenyl chloride, benzenesulfenyl chloride, 2-nitro, o-nitrobenzenesulfenyl chloride, o-nitrophenylsulfenyl chloride, 2-nitrobenzenesulfenylchloride, o-nitrophenylsulphenyl chloride, o-nitrobenzenesulphenyl chloride, 2-nitrophenyl thiohypochlorite, 2-nitrophenyl sulfanyl chlorane | |
| C1=CC=C(C(=C1)[N+](=O)[O-])SCl | |
| 189.613 | |
| 189.61 | |
| Crystalline Powder |
| 7669-54-7 | |
| Yellow | |
| MFCD00007128 | |
| 3261 | |
| NTNKNFHIAFDCSJ-UHFFFAOYSA-N | |
| (2-nitrophenyl) thiohypochlorite | |
| 24319 | |
| ≥95.0% (GC,T) |
Chemical Identifiers
| 7669-54-7 | |
| 189.613 | |
| NTNKNFHIAFDCSJ-UHFFFAOYSA-N | |
| 24319 | |
| C1=CC=C(C(=C1)[N+](=O)[O-])SCl |
| C6H4ClNO2S | |
| MFCD00007128 | |
| 2-nitrobenzenesulfenyl chloride, 2-nitrophenylsulfenyl chloride, benzenesulfenyl chloride, 2-nitro, o-nitrobenzenesulfenyl chloride, o-nitrophenylsulfenyl chloride, 2-nitrobenzenesulfenylchloride, o-nitrophenylsulphenyl chloride, o-nitrobenzenesulphenyl chloride, 2-nitrophenyl thiohypochlorite, 2-nitrophenyl sulfanyl chlorane | |
| (2-nitrophenyl) thiohypochlorite |
Safety and Handling
TSCA : Yes