missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrophenylarsonic Acid 98.0+%, TCI America™

Catalog No. N01331G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N01331G 1 g
N013325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N01331G Supplier TCI America Supplier No. N01331G

Chemical Identifiers

CAS 5410-29-7
Molecular Formula C6H6AsNO5
Molecular Weight (g/mol) 247.038
MDL Number MFCD00047664
InChI Key UYEDGVZDGVIURN-UHFFFAOYSA-N
Synonym 2-nitrobenzenearsonic acid, arsonic acid, 2-nitrophenyl, o-nitrophenylarsenic acid, o-nitrobenzenearsonic acid, o-nitrophenyl arsonic acid, benzenearsonic acid, o-nitro, unii-hnr3su2y5p, 2-nitrophenyl arsonic acid, arsonic acid, as-2-nitrophenyl, hnr3su2y5p
PubChem CID 79424
IUPAC Name (2-nitrophenyl)arsonic acid
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])[As](=O)(O)O

Specifications

Quantity 1 g
UN Number 3465
Formula Weight 247.04
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitrophenylarsonic Acid
RTECSNumber CY5950000
TSCA Yes