Learn More
2-Nitrophenylacetonitrile, 98%
CAS: 610-66-2 | C8H6N2O2 | 162.15 g/mol
$150.22 - $494.29
Chemical Identifiers
| CAS | 610-66-2 |
|---|---|
| Molecular Formula | C8H6N2O2 |
| Molecular Weight (g/mol) | 162.15 |
| MDL Number | MFCD00007183 |
| InChI Key | YPRFCQAWSNWRLM-UHFFFAOYSA-N |
| Synonym | 2-nitrophenylacetonitrile, 2-2-nitrophenyl acetonitrile, 2-nitrophenyl acetonitrile, benzeneacetonitrile, 2-nitro, 2-nitrobenzyl cyanide, o-nitrophenyl acetonitrile, o-nitrobenzacetonitrile, 2-nitrobenzeneacetonitrile, 2-nitrobenzyl nitrile, acetonitrile, o-nitrophenyl |
| PubChem CID | 11888 |
| IUPAC Name | 2-(2-nitrophenyl)acetonitrile |
| SMILES | [O-][N+](=O)C1=CC=CC=C1CC#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2019006
|
Thermo Scientific Chemicals
B2019006 |
5 g |
Each for $150.22
|
|
|||||
|
AAB2019014
|
Thermo Scientific Chemicals
B2019014 |
25 g |
Each for $494.29
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 610-66-2 | |
| 162.15 | |
| YPRFCQAWSNWRLM-UHFFFAOYSA-N | |
| 11888 | |
| [O-][N+](=O)C1=CC=CC=C1CC#N |
| C8H6N2O2 | |
| MFCD00007183 | |
| 2-nitrophenylacetonitrile, 2-2-nitrophenyl acetonitrile, 2-nitrophenyl acetonitrile, benzeneacetonitrile, 2-nitro, 2-nitrobenzyl cyanide, o-nitrophenyl acetonitrile, o-nitrobenzacetonitrile, 2-nitrobenzeneacetonitrile, 2-nitrobenzyl nitrile, acetonitrile, o-nitrophenyl | |
| 2-(2-nitrophenyl)acetonitrile |
Specifications
| 610-66-2 | |
| 178°C (12 mmHg) | |
| MFCD00007183 | |
| UN3439 | |
| 2-nitrophenylacetonitrile, 2-2-nitrophenyl acetonitrile, 2-nitrophenyl acetonitrile, benzeneacetonitrile, 2-nitro, 2-nitrobenzyl cyanide, o-nitrophenyl acetonitrile, o-nitrobenzacetonitrile, 2-nitrobenzeneacetonitrile, 2-nitrobenzyl nitrile, acetonitrile, o-nitrophenyl | |
| [O-][N+](=O)C1=CC=CC=C1CC#N | |
| 162.15 | |
| 162.15 | |
| 2-Nitrophenylacetonitrile |
| 81°C to 85°C | |
| C8H6N2O2 | |
| 5 g | |
| 1869400 | |
| YPRFCQAWSNWRLM-UHFFFAOYSA-N | |
| 2-(2-nitrophenyl)acetonitrile | |
| 11888 | |
| 98% |
Safety and Handling
GHS H Statement
H311-H302-H332
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber : 210-231-0
RTECSNumber : AM1248000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only