missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrophenylacetonitrile, 98%

Catalog No. AAB2019014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2019014 25 g
AAB2019006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2019014 Supplier Thermo Scientific Chemicals Supplier No. B2019014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 610-66-2 | C8H6N2O2 | 162.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 610-66-2
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD00007183
InChI Key YPRFCQAWSNWRLM-UHFFFAOYSA-N
Synonym 2-nitrophenylacetonitrile, 2-2-nitrophenyl acetonitrile, 2-nitrophenyl acetonitrile, benzeneacetonitrile, 2-nitro, 2-nitrobenzyl cyanide, o-nitrophenyl acetonitrile, o-nitrobenzacetonitrile, 2-nitrobenzeneacetonitrile, 2-nitrobenzyl nitrile, acetonitrile, o-nitrophenyl
PubChem CID 11888
IUPAC Name 2-(2-nitrophenyl)acetonitrile
SMILES [O-][N+](=O)C1=CC=CC=C1CC#N

Specifications

Melting Point 81°C to 85°C
Boiling Point 178°C (12 mmHg)
Quantity 25 g
UN Number UN3439
Beilstein 1869400
Formula Weight 162.15
Percent Purity 98%
Chemical Name or Material 2-Nitrophenylacetonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H311-H302-H332
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 210-231-0
RTECSNumber AM1248000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only