missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrophenylacetic Acid 98.0+%, TCI America™

Catalog No. N035325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N035325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N035325G Supplier TCI America Supplier No. N035325G

Chemical Identifiers

CAS 3740-52-1
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.15
MDL Number MFCD00007190
InChI Key WMUZDBZPDLHUMW-UHFFFAOYSA-N
Synonym 2-nitrophenylacetic acid, 2-2-nitrophenyl acetic acid, 2-nitrophenyl acetic acid, o-nitrophenylacetic acid, benzeneacetic acid, 2-nitro, acetic acid, o-nitrophenyl, ortho-nitrophenyl acetic acid, 2-o-nitrophenyl acetic acid, 2-nitrobenzeneacetic acid, unii-05tn0suy38
PubChem CID 77337
IUPAC Name 2-(2-nitrophenyl)acetic acid
SMILES OC(=O)CC1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 141°C
Color Yellow
Quantity 25 g
Formula Weight 181.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitrophenylacetic Acid
RTECSNumber AJ1130000
TSCA Yes