missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrophenylacetic acid, 99%

Catalog No. AAA1341122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1341122 100 g
AAA1341114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1341122 Supplier Thermo Scientific Chemicals Supplier No. A1341122

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3740-52-1 | C8H7NO4 | 181.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3740-52-1
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.15
MDL Number MFCD00007190
InChI Key WMUZDBZPDLHUMW-UHFFFAOYSA-N
Synonym 2-nitrophenylacetic acid, 2-2-nitrophenyl acetic acid, 2-nitrophenyl acetic acid, o-nitrophenylacetic acid, benzeneacetic acid, 2-nitro, acetic acid, o-nitrophenyl, ortho-nitrophenyl acetic acid, 2-o-nitrophenyl acetic acid, 2-nitrobenzeneacetic acid, unii-05tn0suy38
PubChem CID 77337
IUPAC Name 2-(2-nitrophenyl)acetic acid
SMILES OC(=O)CC1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 139°C to 142°C
Odor Odorless
Quantity 100 g
Beilstein 1959243
Formula Weight 181.15
Percent Purity 99%
Chemical Name or Material 2-Nitrophenylacetic acid
Hazard Category H315-H319-H335-H341
Hazard Statement GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
EINECSNumber 223-128-0
RTECSNumber AJ1130000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only