missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrophenyl Phenyl Sulfide 98.0+%, TCI America™

Catalog No. N07445G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N07445G 5 g
N074425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N07445G Supplier TCI America Supplier No. N07445G
Only null left

Chemical Identifiers

CAS 4171-83-9
Molecular Formula C12H9NO2S
Molecular Weight (g/mol) 231.269
InChI Key ZPWNCSAEXUDWTN-UHFFFAOYSA-N
Synonym 2-Nitrodiphenyl Sulfide, 1-Nitro-2-(phenylthio)benzene
PubChem CID 348620
IUPAC Name 1-nitro-2-phenylsulfanylbenzene
SMILES C1=CC=C(C=C1)SC2=CC=CC=C2[N+](=O)[O-]

Specifications

Melting Point 82°C
Color Yellow
Quantity 5 g
Formula Weight 231.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitrophenyl Phenyl Sulfide