Learn More
2-Nitrophenyl isothiocyanate, 97%
CAS: 2719-30-4 | C7H4N2O2S | 180.181 g/mol
Supplier: Thermo Scientific Chemicals L0966906
Description
2-Nitrophenyl isothiocyanate can react with pyridin-2-ylamine to get 2-nitrophenyl(2-pyridyl)thiourea. It is also used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Nitrophenyl isothiocyanate can react with pyridin-2-ylamine to get 2-nitrophenyl(2-pyridyl)thiourea. It is also used as pharmaceutical intermediates.
Solubility
Hydrolyzes with water.
Notes
Store
Specifications
| 2-Nitrophenyl isothiocyanate | |
| 73°C to 76°C | |
| >110°C (230°F) | |
| C7H4N2O2S | |
| 5 g | |
| 2719657 | |
| 2-nitrophenyl isothiocyanate, benzene, isothiocyanatonitro, 2-nitrobenzenisothiocyanate, 2-nitrophenylisothiocyanate, o-nitrophenyl isothiocyanate, 1-isothiocyanato-2-nitro-benzene, # | |
| CBWJHIXSVFDERH-UHFFFAOYSA-N | |
| 1-isothiocyanato-2-nitrobenzene | |
| 137690 | |
| 97% |
| 2719-30-4 | |
| 135°C to 137°C (17 mmHg) | |
| Irritating | |
| MFCD00007092 | |
| UN2923 | |
| Moisture sensitive | |
| Hydrolyzes with water. | |
| C1=CC=C(C(=C1)N=C=S)[N+](=O)[O-] | |
| 180.181 | |
| 180.19 |
Chemical Identifiers
| 2719-30-4 | |
| 180.181 | |
| CBWJHIXSVFDERH-UHFFFAOYSA-N | |
| 137690 | |
| C1=CC=C(C(=C1)N=C=S)[N+](=O)[O-] |
| C7H4N2O2S | |
| MFCD00007092 | |
| 2-nitrophenyl isothiocyanate, benzene, isothiocyanatonitro, 2-nitrobenzenisothiocyanate, 2-nitrophenylisothiocyanate, o-nitrophenyl isothiocyanate, 1-isothiocyanato-2-nitro-benzene, # | |
| 1-isothiocyanato-2-nitrobenzene |
Safety and Handling
GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only