missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Nitrophenoxyacetic Acid 98.0+%, TCI America™
$130.48 - $441.78
Chemical Identifiers
| CAS | 1878-87-1 |
|---|---|
| Molecular Formula | C8H7NO5 |
| Molecular Weight (g/mol) | 197.146 |
| MDL Number | MFCD00016993 |
| InChI Key | TYHHDWAHJRRYCU-UHFFFAOYSA-N |
| Synonym | 2-nitrophenoxyacetic acid, 2-nitrophenoxy acetic acid, 2-2-nitrophenoxy acetic acid, o-nitrophenoxyacetic acid, acetic acid, 2-nitrophenoxy, o-nitrophenoxy acetic acid, acmc-209eqj, 2-nitrophenoxyaceticacid |
| PubChem CID | 15883 |
| ChEBI | CHEBI:66923 |
| SMILES | C1=CC=C(C(=C1)[N+](=O)[O-])OCC(=O)O |
Chemical Identifiers
| 1878-87-1 | |
| 197.146 | |
| TYHHDWAHJRRYCU-UHFFFAOYSA-N | |
| 15883 | |
| C1=CC=C(C(=C1)[N+](=O)[O-])OCC(=O)O |
| C8H7NO5 | |
| MFCD00016993 | |
| 2-nitrophenoxyacetic acid, 2-nitrophenoxy acetic acid, 2-2-nitrophenoxy acetic acid, o-nitrophenoxyacetic acid, acetic acid, 2-nitrophenoxy, o-nitrophenoxy acetic acid, acmc-209eqj, 2-nitrophenoxyaceticacid | |
| CHEBI:66923 |
Specifications
| 1878-87-1 | |
| Yellow | |
| MFCD00016993 | |
| 2-nitrophenoxyacetic acid, 2-nitrophenoxy acetic acid, 2-2-nitrophenoxy acetic acid, o-nitrophenoxyacetic acid, acetic acid, 2-nitrophenoxy, o-nitrophenoxy acetic acid, acmc-209eqj, 2-nitrophenoxyaceticacid | |
| C1=CC=C(C(=C1)[N+](=O)[O-])OCC(=O)O | |
| 15883 | |
| 197.15 | |
| Crystalline Powder |
| 157°C | |
| C8H7NO5 | |
| 5 g | |
| TYHHDWAHJRRYCU-UHFFFAOYSA-N | |
| 197.146 | |
| CHEBI:66923 | |
| ≥98.0% (GC,T) | |
| 2-Nitrophenoxyacetic Acid |
Safety and Handling
TSCA : No