missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrophenoxyacetic Acid 98.0+%, TCI America™

Catalog No. N02255G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N02255G 5 g
N022525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N02255G Supplier TCI America Supplier No. N02255G
Only null left

Chemical Identifiers

CAS 1878-87-1
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.146
MDL Number MFCD00016993
InChI Key TYHHDWAHJRRYCU-UHFFFAOYSA-N
Synonym 2-nitrophenoxyacetic acid, 2-nitrophenoxy acetic acid, 2-2-nitrophenoxy acetic acid, o-nitrophenoxyacetic acid, acetic acid, 2-nitrophenoxy, o-nitrophenoxy acetic acid, acmc-209eqj, 2-nitrophenoxyaceticacid
PubChem CID 15883
ChEBI CHEBI:66923
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])OCC(=O)O

Specifications

Melting Point 157°C
Color Yellow
Quantity 5 g
Formula Weight 197.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitrophenoxyacetic Acid