missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrofluorenone 98.0+%, TCI America™

Catalog No. N02021G
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N02021G 1 g
N020210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N02021G Supplier TCI America Supplier No. N02021G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3096-52-4
Molecular Formula C13H7NO3
Molecular Weight (g/mol) 225.20
MDL Number MFCD00001152
InChI Key AJEAHBZZHSLIQP-UHFFFAOYSA-N
Synonym 2-nitro-9h-fluoren-9-one, 2-nitro-9-fluorenone, 2-nitrofluorenone, 9h-fluoren-9-one, 2-nitro, ccris 2540, 9-fluorenone, 2-nitro, 4-07-00-01636 beilstein handbook reference, 2-nitro-9h-fluoren-9-one #
PubChem CID 18356
IUPAC Name 2-nitro-9H-fluoren-9-one
SMILES [O-][N+](=O)C1=CC=C2C3=CC=CC=C3C(=O)C2=C1

Specifications

Melting Point 222°C
Color Yellow
Quantity 1 g
Formula Weight 225.20
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitrofluorenone
RTECSNumber LL9091000
TSCA No