missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroethanol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 397960250

 View more versions of this product

Catalog No. AC397960250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Nitroethanol
96.0
1.2920g/mL
>112°C
96% min. (GC)
C2H5NO3
O2NCH2CH2OH
25g
ethanol, 2-nitro, 2-nitroethan-1-ol, beta-nitroalcohol, beta-nitroethanol, 2-nitro-ethanol, .beta.-nitroethanol, ccris 6076, 2 nitroethanol, pubchem14208, 2-nitroethanol
KIPMDPDAFINLIV-UHFFFAOYSA-N
2-nitroethanol
12252
97%
625-48-9
100.0
203.0°C to 206.0°C
Authentic
Glass bottle
1.4445
MFCD00007405
1.292
Solubility in water: miscible
C(CO)[N+](=O)[O-]
91.07
91.07

Chemical Identifiers

625-48-9
91.07
KIPMDPDAFINLIV-UHFFFAOYSA-N
12252
C(CO)[N+](=O)[O-]
C2H5NO3
MFCD00007405
ethanol, 2-nitro, 2-nitroethan-1-ol, beta-nitroalcohol, beta-nitroethanol, 2-nitro-ethanol, .beta.-nitroethanol, ccris 6076, 2 nitroethanol, pubchem14208, 2-nitroethanol
2-nitroethanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-895-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only